Ammoniakkiliuos 14%, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Ammoniakki

0889-15EA 0 EUR
0889-15
Ammoniakkiliuos 14%, J.T.Baker®
Ammoniakkiliuos
Kaava: NH₃
MW: 35.05 g/mol
MDL numero: MFCD00066650
CAS-numero: 1336-21-6
UN: 2672
ADR: 8 III

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (as NH₃) 11.2 - 16.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR