Polysorbaatti 20, Reagent Grade

Toimittaja: VWR Chemicals

0777-1LEA 82.5 EUR
0777-1L 0777-4L
Polysorbaatti 20, Reagent Grade
Polysorbaatti 20

Non-ionic detergent for solubilising membrane proteins without altering biological activity.

Kaava: C₅₈H₁₁₄O₂₆
Kiehumispiste: >100 °C
Tiheys: 1,1 g/cm³ (20 °C)
Leimahduspiste: 149 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00165986
CAS-numero: 9005-64-5
EINECS: 500-018-3

Order Now

Testitulosten erittely

Arsenic <0.0003%
DNase NONE
Heavy Metals <0.001%
Hydroxyl Number 96 – 108
Identification PASS
Moisture (KF) 3.0%
Protease NONE
Residue on Ignition 0.25%
RNase NONE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR