Ammoniumpersulfaatti (APS) ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

0762.1000EA 81.52 EUR
0762.1000 0762.5000 0762-07
Ammoniumpersulfaatti (APS) ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumpersulfaatti (APS)

Varoitus: Because of inherent instability, this reagent may be expected to decrease in strength and to increase in acidity during storage.

Kaava: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Sulamispiste: 120
Tiheys: 1.98 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003390
CAS-numero: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1 III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Meets ACS Reagent Chemical Requirements ≥98.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
Store below 20°C. ≤0.050%
Assay ((NH₄)₂S₂O₈) ≥ 98.0 %
Insoluble Matter ≤ 0.005 %
Residue after Ignition ≤ 0.050 %
Titrable Free Acid (meq/g) ≤ 0.04
Chloride and Chlorate (as Cl) ≤ 0.001 %
Heavy Metals (as Pb) ≤ 0.005 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
Trace Impurities - Manganese (Mn) ≤ 0.500 ppm
Because of inherent instability, this reagent may be expected to
decrease in strength and to increase in acidity during storage.
After storage for some time, the reagent may fail to meet the specified
requirements for Assay and Titrable Free Acid.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR