Kloorivety liuos 0.5 M metanolissa, LiChropur™ for GC derivatisation, Supelco®

Toimittaja: Merck
Vaara

07607-10X1ML 07607-100ML
07607-10X1MLEA 61.4 EUR
07607-10X1ML 07607-100ML
Kloorivety liuos 0.5 M metanolissa, LiChropur™ for GC derivatisation, Supelco®
Kloorivety

Methanolic HCl was used in fatty acid analysis during a study to identify Δ4 fatty acid desaturated from Thraustochytrium sp. involved in the biosynthesis of docosahexanoic acid.

Kaava: HCl
Tiheys: 0,800...0.811 g/m³ (20 °C)
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Testitulosten erittely

Appearance (Color) Colorless
Appearance (Form) Liquid
Titration (Method) Alkalimetric
Titration (Molarity) 0.45 - 0.55 mol/l
Remarks on GC Suitibility for GC derivatization complying
Density D20/4 0.800 - 0.811
Proton NMR Spectrum Conforms to Structure
Recommended Retest Period 54 Months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR