Imidatsoli ≥99% for biotechnology

Toimittaja: VWR Chemicals
Vaara

0527-100GEA 232 EUR
0527-100G 0527-50G 0527-1KG
Imidatsoli ≥99% for biotechnology
Imidatsoli

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological.

Kaava: C₃H₄N₂
Kiehumispiste: 255 °C (1013 hPa)
Sulamispiste: 89…90 °C
Tiheys: 1,18 g/cm³ (20 °C)
Leimahduspiste: 135 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005183
CAS-numero: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8 (6.1),III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

DNase none detected
Insolubles ≤0.1%
Melting Point 88 – 92 °C
pH (5%, Water) @25 °C 9.5 – 10.5
Protease none
Purity >99%
RNase none detected
Solubility (20%, Water) pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR