Dinatriumvetyfosfaatti-7 H2O ACS

Toimittaja: VWR Chemicals

0348-1KGEA 132 EUR
0348-1KG 0348-2.5KG 0348-500G 0348-12KG 0348-50KG
Dinatriumvetyfosfaatti-7 H2O ACS
Dinatriumvetyfosfaatti-7 H2O
Kaava: HNa₂O₄P·7H₂O
Sulamispiste: 48 °C
Tiheys: 1,7 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149180
CAS-numero: 7782-85-6
EINECS: 231-448-7

Order Now

Testitulosten erittely

Chloride <0.001%
Heavy Metals (as Pb) ≤0.001%
Insolubles ≤0.005%
Iron ≤0.001%
pH (5%, Water) at 25 °C 8.7 – 9.3
Purity (as is) 98 – 102%
Sulphate ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR