D(+)-sakkaroosi ≥99.9%, Ultrapure DNase-, RNase-free

Toimittaja: VWR Chemicals

0335-1KGEA 92 EUR
0335-1KG 0335-5KG 0335-500G 0335-10KG 0335-12KG 0335-2.5KG 0335-50KG
D(+)-sakkaroosi ≥99.9%, Ultrapure DNase-, RNase-free
D(+)-sakkaroosi

DNase- and RNase-free. Common reagent in routine nucleic acid procedures and electrophoresis loading dyes.

Kaava: C₁₂H₂₂O₁₁
Sulamispiste: 169…170 °C
Tiheys: 1,29 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006626
CAS-numero: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Chloride ≤0.005%
DNase None detected
Glucose ≤0.1%
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.005%
Invert Sugar ≤0.1%
Iron ≤0.0005%
Loss on Drying ≤0.03%
Protease none detected
Purity ≤99.9%
RNase none detected
Solubility (10%, Water) Pass
Specific Rotation 66.3 – 66.8°
Sulphate ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR