EDTA free acid ≥99.5%, Ultrapure

Toimittaja: VWR Chemicals
Varoitus

0322-1KGEA 510 EUR
0322-1KG 0322-500G
EDTA free acid ≥99.5%, Ultrapure
EDTA

Calcium chelator used to eliminate inhibition of enzyme catalysed reactions due to traces of heavy metals.

Kaava: C₁₀H₁₆N₂O₈
Kiehumispiste: 614 °C (1013 hPa)
Sulamispiste: 220 °C
Tiheys: 0,86 g/cm³ (20 °C)
Leimahduspiste: >100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003541
CAS-numero: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

DNase none detected
Heavy Metals (as Pb) ≤0.001%
Insolubles (5%, 1N NaOH) Pass
Iron ≤0.005%
Magnesium ≤0.0005%
Nitrilotriacetic Acid ≤0.1%
pH (5%, Water) @25 °C 2.5 – 3.5
Protease none detected
Purity ≥99.5%
Residue after Ignition ≤0.2%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR