Kaliumjodidi 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

Synonyms: Dipotassium diiodide, Kali iodide

03124-1KG 03124-250G 03124-5KG 03124-6X1KG 03124-6X250G
03124-1KGEA 325.49 EUR
03124-1KG 03124-250G 03124-5KG 03124-6X1KG 03124-6X250G
Kaliumjodidi 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Kaliumjodidi
Kaava: Kl
MW: 166 g/mol
Kiehumispiste: 1.323 °C (1.013 hPa)
Sulamispiste: 681 °C
Tiheys: 3.12 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011405
CAS-numero: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Testitulosten erittely

Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying (105°C) Max. 0.5 %
Alkalinely reacting substances complying
Iron (Fe) Max. 10 ppm
Heavy metals (as Pb) Max. 10 ppm
Iodate (JO₃) complying
Iodate (IO₃) Max. 0.0004 %
Sulfate (SO₄) Max. 100 ppm
Thiosulfate (S₂O₃) Max. 0.01 %
Nitrate, nitrite, ammonium complying
Thiosulfate (S₂O₃) and Ba complying
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR