Tetranatriumdifosfaatti-10 H2O, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Varoitus

0309.1000EA 90.99 EUR
0309.1000 0309.9050
Tetranatriumdifosfaatti-10 H2O, BAKER ANALYZED® ACS, J.T.Baker®
Tetranatriumdifosfaatti-10 H2O
Kaava: Na₄P₂O₇·10H₂O
Sulamispiste: 80 °C
Tiheys: 1,82 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149200
CAS-numero: 13472-36-1
EINECS: 231-767-1

Order Now

Testitulosten erittely

Meets ACS Specifications
Assay 99.0 - 103.0 %
Chloride (Cl) max. 0.002 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.01 %
Nitrogen Compounds (as N) max. 0.001 %
pH of 5% Solution at 25°C 9.5 - 10.5
Sulfate (SO4) max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR