VWR® UltraPure, Tris-glycine buffers, 10X concentrate
Toimittaja: VWR Chemicals
0307-4LEA 278 EUR
0307-4L 0307-1L 0307-5L
VWR® UltraPure, Tris-glycine buffers, 10X concentrate
Biokemialliset puskuriliuokset Elektroforeesipuskurit Running Buffers
Running buffers for both 1-D and 2-D protein gel electrophoresis.
  • Concentrated solutions with a choice of packaging options.
A 1X working solution contains 0,025 M Tris base and 0,192 M glycine.