Natriumvetykarbonaatti, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor

Synonyms: Sodium bicarbonate

0263.0500EA 28.1 EUR
0263.0500 0263.1000 0263.5000
Natriumvetykarbonaatti, BAKER ANALYZED® ACS, J.T.Baker®
Natriumvetykarbonaatti
Kaava: NaHCO₃
MW: 84.01 g/mol
Sulamispiste: 270
Tiheys: 2.159 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003528
CAS-numero: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Meets ACS Specifications
Assay (dried basis) 99.7 - 100.3 %
Calcium (Ca) max. 0.02 %
Chloride (Cl) max. 0.003 %
Iron (Fe) max. 0.001 %
Potassium (K) max. 0.005 %
Magnesium (Mg) max. 0.005 %
Insoluble Matter max. 0.015 %
Phosphate (PO4) max. 0.001 %
Sulfur Compounds (as SO4) max. 0.003 %
Trace Impurities (in ppm)
Ammonium (NH4) max. 5
Heavy Metals (as Pb) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR