Fenoli, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

0188.0500EA 113.44 EUR
0188.0500 0188.1000
Fenoli, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Fenoli
Kaava: C₆H₅OH
MW: 94.11 g/mol
Kiehumispiste: 181.7
Sulamispiste: 40.8
Tiheys: 1.06 g/cm³ (20 °C)
Leimahduspiste: 81 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002143
CAS-numero: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Order Now

Testitulosten erittely

Exceeds ACS Specifications
Assay (by GC) min. 99.0 %
Clarity of Solution passes test
Freezing Point min. 40.5 °C
Residue after Evaporation max. 0.03 %
Water (H2O) max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR