VWR® Acrylamide and Bis-Acrylamide
Toimittaja: VWR Chemicals
0172-250GEA 299 EUR
0172-250G 0172-50G 0254-500ML 0172-100G
VWR® Acrylamide and Bis-Acrylamide
Elektroforeesigeelit Agaroosi ja akryyliamidi
An extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids.
Ultra-pure (>99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications.
Liquid blends minimise the handling of neurotoxic acrylamide.
Pakkaus: Packed in a plastic bottle.