Sitruunahappo monohydraatti 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®

Toimittaja: Avantor
Varoitus

0115-01EA 111.53 EUR
0115-01 0115-05 0115-07 0115-09 0115-32 0115-34 0115-37 0115-39
Sitruunahappo monohydraatti 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Sitruunahappomonohydraatii

Varoitus: Must be subjected to further processing during the preparation of injectable dosage forms.

Kaava: C₆H₈O₇.H₂O
Sulamispiste: 135…152 °C
Tiheys: 1,54 g/cm³ (20 °C)
Leimahduspiste: 173,9 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149972
CAS-numero: 5949-29-1
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Meets E.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification Passes Test
USP - Water (H₂O) 7.5 - 9.0 %
USP - Readily Carbonizable Substances Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Sulfate (SO₄) ≤0.015 %
USP - Oxalic Acid ≤0.036 %
USP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
EP/BP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Readily Carbonizable Substances Passes Test
EP/BP - Oxalic Acid ≤360 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Water (H₂O) 7.5 - 9.0 %
EP/BP - Ash (sulfated) ≤0.1 %
EP/BP - Endotoxin Concentration, IU/mg ≤0.5
JP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
JP - Identification Passes Test
JP - Sulfate (SO₄) ≤150 ppm
JP - Oxalic Acid ≤360 ppm
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Water (H₂O) 7.5 - 9.0 %
JP - Readily Carbonizable Substances Passes Test
JP - Residue on Ignition ≤0.10 %
Must be subjected to further processing during the preparation of
injectable dosage forms.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR