Magnesium ≥99.8% (metals basis), granules -12+50 mesh

Toimittaja: Thermo Fisher Scientific
Vaara

00870.36 00870.A1 00870.A3
00870.36EA 261 EUR
00870.36 00870.A1 00870.A3
Magnesium ≥99.8% (metals basis), granules -12+50 mesh
Magnesium

Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver-grey Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Kaava: Mg
MW: 24.3 g/mol
Kiehumispiste: 1107
Sulamispiste: 651
Tiheys: 1.74 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00085308
CAS-numero: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR