Litium 99% (metals basis), rakeita 1-6 mm (0.04-0.2 in)

Toimittaja: Thermo Fisher Scientific
Vaara


Packaged under argon.

00733.18 00733.30
00733.18EA 390 EUR
00733.18 00733.30
Litium 99% (metals basis), rakeita 1-6 mm (0.04-0.2 in)
Litium

Purity: 99% CAS No.: 7439-93-2 Physical Form: Packaged under argon Appearance: Silvery Odour: Odourless UN No.: 1415 MDL No.: MFCD00134051

Kaava: Li
Kiehumispiste: 1317 °C (1013 hPa)
Sulamispiste: 179 °C
Tiheys: 0,53 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134051
CAS-numero: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3,I
Merck Index: 13,05542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR