Nikkeli ≥99.8% (metals basis), spherical powder, APS 5-15 µm

Toimittaja: Thermo Fisher Scientific
Vaara

00663.14 00663.22
00663.14EA 177 EUR
00663.14 00663.22
Nikkeli ≥99.8% (metals basis), spherical powder, APS 5-15 µm
Nikkeli

Purity: 99.8% CAS No.: 7440-02-0 Physical Form: Apparent density 3-5g/cm3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011137

Kaava: Ni
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011137
CAS-numero: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR