Kupari ≥99% (metals basis), jauhe -625 mesh (APS 2-3,50 µm)

Toimittaja: Thermo Fisher Scientific
Varoitus


Packaged under argon.

00622.09 00622.18 00622.30
00622.09EA 98.7 EUR
00622.09 00622.18 00622.30
Kupari ≥99% (metals basis), jauhe -625 mesh (APS 2-3,50 µm)
Kupari

Purity: 99% CAS No.: 7440-50-8 Physical Form: Packaged under argon Appearance: Reddish Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965

Kaava: Cu
Kiehumispiste: 2595 °C (1013 hPa)
Sulamispiste: 1083 °C
Tiheys: 8,94 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010965
CAS-numero: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR