Ammoniumsulfaatti, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

0049.9050EA 731.96 EUR
0049.9050 0049.9025BB 0049.5000 0049.1000
Ammoniumsulfaatti, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumsulfaatti
Kaava: (NH₄)₂SO₄
MW: 132.14 g/mol
Sulamispiste: 336…339 °C
Tiheys: 1.78 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003391
CAS-numero: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Testitulosten erittely

Meets ACS Specifications
Assay min. 99.0 %
Insoluble Matter max. 0.005 %
Nitrate (NO3) max. 0.001 %
pH of 5% Solution at 25°C 5.0 - 6.0
Residue after Ignition max. 0.005 %
Trace Impurities (in ppm)Chloride (Cl) max. 5
Trace Impurities (in ppm)Iron (Fe) max. 5
Trace Impurities (in ppm)Heavy Metals (as Pb) max. 5
Trace Impurities (in ppm)Phosphate (PO4) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR