Indium ≥99.999% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)

Toimittaja: Thermo Fisher Scientific

00155.G2 00155.0K
00155.G2EA 582 EUR
00155.G2 00155.0K
Indium ≥99.999% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Indium

Purity: 99.999% CAS No.: 7440-74-6 Physical Form: ≈1.43g/m Odour: Odourless MDL No.: MFCD00134048

Kaava: In
MW: 114.82 g/mol
Kiehumispiste: 2000
Sulamispiste: 156
Tiheys: 7.3 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134048
CAS-numero: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR