Anti-Leukotriene B4 Receptor 1 Rabbit Polyclonal Antibody

Toimittaja: US Biological
136225
USBI136225EA 632 EUR
USBI136225
Anti-Leukotriene B4 Receptor 1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended dilution: Western Blot: 1:200; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Leukotriene B4 Receptor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR