Isopentylamine hydrobromide ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

I1094-1G I1094-5G
TCIAI1094-1GEA 30.8 EUR
TCIAI1094-1G TCIAI1094-5G
Isopentylamine hydrobromide ≥98.0% (by titrimetric analysis)
Isopentylamine hydrobromide
Kaava: C₅H₁₃N·HBr
MDL numero: MFCD33021450
CAS-numero: 2733412-57-0

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR