Nipagin A, etyyli-4-hydroksibentsoaatti ≥99.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

H0211-25G H0211-500G
TCIAH0211-25GEA 17.6 EUR
TCIAH0211-25G TCIAH0211-500G
Nipagin A, etyyli-4-hydroksibentsoaatti ≥99.0% (by GC, titration analysis)
Nipagin A, etyyli-4-hydroksibentsoaatti
Kaava: C₉H₁₀O₃
Kiehumispiste: 178°C
Sulamispiste: 116…118 °C
Tiheys: 1,078 g/cm³ (131 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002353
CAS-numero: 120-47-8
EINECS: 204-399-4
Merck Index: 14,03837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff9fa1b430130b67976c849d1e016d806c918cbf&vid=d92b68bc82745b396d5edc9bd40b831b3da16a34&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 115.0 to 118.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR