Pullo, kapea suuaukko, diagnostiset pullot, pyöreät, Nalgene®
Toimittaja: Thermo Fisher Scientific
362008-9125 362008-9025 362008-9050
NALG362008-9125EA 1880 EUR
NALG362008-9125 NALG362008-9025 NALG362008-9050
Pullo, kapea suuaukko, diagnostiset pullot, pyöreät, Nalgene®
Pullot Muovipullot
Opaque bottles without closure, used to store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples.
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof when assembled with Nalgene closure
Sertifikaatit: Compliance with ISO 13485:2003 and 21CFR177.1520, USP Class VI.
Tilaustiedot: Coloured closures are ordered seperately.