McMillan Flow Modular Liquid Flow Rate Sensors
Toimittaja: Avantor Fluid Handling
32704-00 32704-02 32704-04 32704-10
MFLX32704-00EA 590 EUR
MFLX32704-00 MFLX32704-02 MFLX32704-04 MFLX32704-10
McMillan Flow Modular Liquid Flow Rate Sensors
Virtausmittarit
System design is flexible to suit a wide range of applications.
  • Brass sensors for high pressure; PTFE sensors for high-purity systems; Ryton® PPS sensors for aggressive liquids
  • Interface with data acquisition systems
  • Dual signal outputs to monitor and record flow rates and totals
Use these modular flow sensors and totalizers for any type of process or laboratory application. The flow sensors measure low viscosity liquids from 13 ml/min up to 10 L/min. For a complete system, order a flow sensor, power supply, and flow display/totalizer.
These sensorsMore Product Information