L(+)-kysteiini, white powder cell culture reagent

Toimittaja: MP Biomedicals
Varoitus

0219464691 0219464625 0219464680 0219464690
ICNA0219464691EA 838 EUR
ICNA0219464691 ICNA0219464625 ICNA0219464680 ICNA0219464690
L(+)-kysteiini, white powder cell culture reagent
L-kysteiini

Storage: Store at Room Temperature (15-30 °C). Store Desiccated. Store under nitrogen.
L-Cysteine is a proteinogenic amino acid incorporated into proteins as directed by the genetic code. The thiol-side chain participates in a variety of oxidation/reduction reactions within the cell. The side chain participates in the formation of β bonds that modulate the secondary and ternary structure of proteins. Cysteine is essential and limiting for the formation of glutathione, an important antioxidant, within the cell.
Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
NMDA glutamatergic receptor agonist.


  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in water: 25 mg/mL - Clear, Colorless Solution.

Kaava: HSCH₂CH(NH₂)CO₂H
Kiehumispiste: 254 °C (1013 hPa)
Sulamispiste: 200…235 °C (haj. lämpötila)
Tiheys: 1,523 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064306
CAS-numero: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810

Order Now

Testitulosten erittely

Identity Test Passes
FTIR Conforms to Standard
Optical Rotation (c=2, 1N HCl)
7 - 13° pH (5% aq soln)
4-8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR