Dekantterit, mittadekantterit, kahvallinen
Toimittaja: HIRSCHMANN
7600394 7600393 7600390 7600386 7600380 7600375
HIRS7600394EA 83 EUR
HIRS7600394 HIRS7600393 HIRS7600390 HIRS7600386 HIRS7600380 HIRS7600375
Dekantterit, mittadekantterit, kahvallinen
Dekantterit
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale, blue graduations.
Food-safe products acc. to EC Directive No. 10/2011.