Anti-EIF2AK3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BS-3330R-A488
BOSSBS-3330R-A488EA 921 EUR
BOSSBS-3330R-A488
Anti-EIF2AK3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Phosphorylates the alpha subunit of eukaryotic translation-initiation factor 2 (EIF2), leading to its inactivation and thus to a rapid reduction of translational initiation and repression of global protein synthesis. Serves as a critical effector of unfolded protein response (UPR)-induced G1 growth arrest due to the loss of cyclin-D1 (CCND1). Involved in control of mitochondrial morphology and function.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: EIF2AK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR