Glyseriini ≥86-88% (w/w) vesiliuos for analysis

Toimittaja: Thermo Fisher Scientific

295600010 295600025 295602500 295600100
ACRO295600010EA 119 EUR
ACRO295600010 ACRO295600025 ACRO295602500 ACRO295600100
Glyseriini ≥86-88% (w/w) vesiliuos for analysis
Glyseriini
Kaava: HOCH₂CH(OH)CH₂OH
Sulamispiste: 18 °C
Tiheys: 1,227 g/cm³
Leimahduspiste: 160 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004722
CAS-numero: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4550 (20°C, 589 nm)
Titration Iodometric 86.0 to 88.0 %
Water 12.0 to 14.0 % (K.F.)
Sulfated ash ≤0.005 %
Density (20/4°C) 1.224 to 1.232 g/ml
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤1 ppm
Sulfate (SO4) ≤5 ppm
Ammonium (NH4) ≤5 ppm
Arsenic (As) ≤0.5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤1 ppm
Free acid (CH3COOH) ≤0.002 %
Free alkali (NH3) ≤0.0005 %
Reducing substances ≤0.003 %
Fatty acid esters ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR