Basic Violet 3, pure indicator

Toimittaja: Thermo Fisher Scientific
Vaara

212120250 212121000 212125000
ACRO212120250EA 39.6 EUR
ACRO212120250 ACRO212121000 ACRO212125000
Basic Violet 3, pure indicator
Basic Violet 3
Kaava: C₂₅H₃₀ClN₃
Sulamispiste: 173 °C (haj. lämpötila)
Tiheys: 1,19 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011750
CAS-numero: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406

Order Now

Testitulosten erittely

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Sensitivity (as indicator) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR