Supelco® TraceCERT® Organic Reference Standard, Isocarbophos
Toimittaja: Merck
36831-50MG
36831-50MGEA 89.1 EUR
36831-50MG
Supelco® TraceCERT® Organic Reference Standard, Isocarbophos
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P264 - P270 - P280 - P301 + P310 - P302 + P352 + P312 - P361 + P364