Bromivety 48% (w/w) vesiliuos

Toimittaja: Thermo Fisher Scientific
Vaara

14036.ATEA 163 EUR
14036.AT 14036.AZ
Bromivety 48% (w/w) vesiliuos
Bromivety

CAS No.: 10035-10-6 Molecular Formula: HBr Formula Weight: 80.92 Physical Form: Liquid Density: 1.490 Sensitivity: Air Sensitive Appearance: Colourless to light yellow Beilstein Reference: 3587158 Merck Reference: 14,4778 UN No.: 1788 MDL No.: MFCD00011323

Kaava: HBr
Tiheys: 1,490
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011323
CAS-numero: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II
Merck Index: 14,4778

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR